Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GS1-120K12.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508903.1 | - | 2 | 998 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566476.1 | - | 2 | 998 | Chr. 1 - 185001527 - 185008683 [+] on Build GRCh38 |