Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134923.1 | 1 | 121 | |||
GenBank mRNA | DB447763.1 | - | 1 | 121 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566649.1 | - | 1 | 87 | Chr. 16 - 54366007 - 54370665 [-] on Build GRCh38 |