Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525928.1 | XP_011524230.1 | 3 | 1159 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000579278.1 | - | 2 | 596 | Chr. 18 - 75432703 - 75436957 [-] on Build GRCh38 |