Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002838.1 | 6 | 1133 | |||
GenBank mRNA | AY762618.1 | - | 6 | 1133 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000592302.5 | - | 6 | 1133 | Chr. 18 - 41480274 - 41520597 [-] on Build GRCh38 |