Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205273.2 | NP_001192202.1 | 2 | 669 | ||
NM_001278575.1 | NP_001265504.1 | 3 | 1079 | |||
NM_001278576.1 | NP_001265505.1 | 4 | 1203 | |||
NM_001278577.1 | NP_001265506.1 | 4 | 1214 | |||
NM_001278578.1 | NP_001265507.1 | 3 | 1072 | |||
XM_011526302.2 | XP_011524604.1 | 3 | 2858 | |||
XM_011526303.2 | XP_011524605.1 | 3 | 1683 | |||
XM_017026126.1 | XP_016881615.1 | 3 | 3194 | |||
XM_017026127.1 | XP_016881616.1 | 3 | 2653 | |||
XM_017026128.1 | XP_016881617.1 | 3 | 2851 | |||
XM_017026129.1 | XP_016881618.1 | 3 | 1690 | |||
XM_017026130.1 | XP_016881619.1 | 3 | 1415 | |||
XM_017026131.1 | XP_016881620.1 | 4 | 1021 | |||
XM_017026132.1 | XP_016881621.1 | 3 | 1214 | |||
XM_017026133.1 | XP_016881622.1 | 2 | 882 | |||
GenBank mRNA | BC043286.1 | - | 4 | 1142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000590611.2 | - | 4 | 1137 | Chr. 19 - 31588255 - 31593781 [+] on Build GRCh38 |