Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259102.4 | XP_005259159.1 | 2 | 99 | ||
XM_017027033.1 | XP_016882522.1 | 2 | 214 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000601911.1 | - | 2 | 90 | Chr. 19 - 39314651 - 39320858 [-] on Build GRCh38 |