Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026592.2 | 12 | 1701 | |||
GenBank mRNA | AK309545.1 | - | 2 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000580790.1 | - | 12 | 1654 | Chr. 10 - 73699151 - 73730487 [-] on Build GRCh38 |