Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111950.1 | 9 | 5407 | |||
GenBank mRNA | AB007962.1 | - | 9 | 5076 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000610578.4 | - | 5075 | Chr. - - [-] on Build |