Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A930002I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080695.1 | - | 2 | 811 | ||
AK088812.1 | - | 2 | 893 | |||
BB277142.2 | - | 1 | 66 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_175410.1 | - | 811 | Chr. - - [-] on Build |