Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002321.2 | 2 | 227 | |||
NR_002322.2 | 1 | 623 | ||||
NR_110490.1 | 1 | 623 | ||||
GenBank mRNA | AK033381.1 | - | 1 | 594 | ||
AK048213.1 | - | 1 | 615 | |||
AK049839.1 | - | 1 | 613 | |||
AK083955.1 | - | 1 | 604 | |||
AK088174.1 | - | 2 | 228 | |||
AK162835.1 | - | 1 | 619 | |||
BC048828.1 | - | 1 | 499 | |||
DQ517434.1 | - | 1 | 623 | |||
DQ517435.1 | - | 2 | 222 |