Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930453O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015456.1 | - | 91 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715004.1 | - | 1, 2 | 91 | Chr. 1 - 88347799 - 88352930 [-] on Build GRCm38 |