Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076389.1 | - | 4 | 2313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC053942.1 | - | 1645 | Chr. - - [-] on Build |