Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem150cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045993.1 | 4 | 419 | |||
GenBank mRNA | AK017893.1 | - | 4 | 420 | ||
AK042097.1 | - | 3 | 452 |