Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem150cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045993.1 | 5 | 1160 | |||
GenBank mRNA | AK017893.1 | - | 5 | 1161 | ||
AK042097.1 | - | 4 | 1194 |