Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm26756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039080.1 | - | 258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632903.1 | - | 2, 3 | 258 | Chr. 5 - 67427680 - 67437243 [+] on Build GRCm38 |