Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094889.1 | - | 1 | 1298 | ||
AK097082.1 | - | 1 | 1865 | |||
BC031001.1 | - | 3 | 928 | |||
EU541207.1 | - | 3 | 874 | |||
M27826.1 | - | 2 | 401 |