siRNA Details
Gene Symbol: C3orf67-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110820.1 | 3 | 2143 | |||
GenBank mRNA | AK090895.1 | - | 3 | 2143 |

Gene Symbol: C3orf67-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110820.1 | 3 | 2143 | |||
GenBank mRNA | AK090895.1 | - | 3 | 2143 |