Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110828.1 | 2 | 1319 | |||
GenBank mRNA | BX648465.1 | - | 2 | 1320 |