Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015353.1 | 4 | 2578 | |||
GenBank mRNA | AK001756.1 | - | 4 | 2578 | ||
AK023798.1 | - | 1 | 3654 | |||
AK129670.1 | - | 1 | 1581 | |||
AK130294.1 | - | 1 | 4781 | |||
AL080202.1 | - | 3 | 2461 | |||
BC024701.1 | - | 1 | 307 | |||
BC048208.1 | - | 1 | 1566 |