Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125926.1 | 6, 7 | 631 | |||
GenBank mRNA | AK094561.1 | - | 6, 7 | 631 |