Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_109974.1 | 4 | 597 | |||
GenBank mRNA | AJ412026.1 | - | LIT1926 BCMS |
3 | 561 | |
AJ412027.1 | - | LIT1925 BCMS |
3 | 268 | ||
AJ412029.1 | - | LIT1923 BCMS |
3 | 268 | ||
AJ412031.1 | - | LIT1917 BCMS |
8 | 1072 | ||
AJ412033.1 | - | LIT1912 BCMS |
2 | 174 | ||
AJ412034.1 | - | LIT1911 BCMS |
2 | 174 | ||
AJ412036.1 | - | LIT1909 BCMS |
2 | 174 | ||
AJ412037.1 | - | LIT1908 BCMS |
2 | 174 | ||
AJ412042.1 | - | LIT1903 BCMS |
3 | 346 | ||
AJ412043.1 | - | LIT1902 BCMS |
4 | 459 | ||
AJ412054.1 | - | LIT1886 BCMS |
5 | 729 | ||
AJ412055.1 | - | LIT1885 BCMS |
4 | 595 | ||
AJ412056.1 | - | LIT1884 BCMS |
4 | 660 | ||
AJ412057.1 | - | LIT1876 BCMS |
5 | 708 | ||
AJ412058.1 | - | LIT1875 BCMS |
4 | 614 | ||
AJ412059.1 | - | LIT1874 BCMS |
5 | 708 | ||
AJ412060.1 | - | LIT1873 BCMS |
3 | 1694 | ||
AJ412061.1 | - | LIT1872 BCMS |
5 | 708 | ||
AJ412062.1 | - | LIT1871 BCMS |
5 | 708 | ||
AJ412063.1 | - | LIT1870 BCMS |
2 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AJ412003.3 | - | 1 | 61 | Chr. 13 - 50433413 - 50433550 [+] on Build GRCh38 |