Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053236.2 | NP_444466.2 | 1 | 153 | ||
GenBank mRNA | BC115501.1 | - | 1 | 153 | 40100944 | |
BC115502.1 | - | 1 | 153 | 40100945 |