Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033521.3 | NP_277056.1 | 5 | 757 | ||
GenBank mRNA | AF317418.1 | - | 5 | 459 | ||
AK077269.1 | - | 5 | 620 | |||
AK077933.1 | - | 5 | 757 | |||
AK079581.1 | - | 5 | 620 | |||
AK082807.1 | - | 5 | 636 | |||
AK146868.1 | - | 4 | 523 | |||
AK166014.1 | - | 5 | 659 | |||
BC019120.1 | - | 5 | 627 | 4507607 | ||
EU561010.1 | - | 5 | 757 |