Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054055.2 | NP_473396.1 | 2 | 295 | ||
GenBank mRNA | AF306491.1 | - | 2 | 294 | ||
AK085326.1 | - | 2 | 293 | |||
AK086775.1 | - | 2 | 297 | |||
AK087518.1 | - | 2 | 293 | |||
AK128982.1 | - | 2 | 292 | |||
AK138344.1 | - | 2 | 293 | |||
AK157881.1 | - | 2 | 264 | |||
AK165355.1 | - | 2 | 293 | |||
BC026803.1 | - | 2 | 281 | 4238769 | ||
BC028882.1 | - | 2 | 279 |