Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053268.2 | NP_444498.2 | 13, 14 | 1374 | ||
GenBank mRNA | AB056433.1 | - | 13, 14 | 1367 | ||
AK034434.1 | - | 13, 14 | 1374 | |||
AK035664.1 | - | 4, 5 | 397 | |||
AK082530.1 | - | 4, 5 | 526 | |||
AK154499.1 | - | 13, 14 | 1391 | |||
AK166654.1 | - | 13, 14 | 1374 |