Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053268.2 | NP_444498.2 | 21 | 2165 | ||
GenBank mRNA | AB056433.1 | - | 21 | 2158 | ||
AK034434.1 | - | 21 | 2165 | |||
AK035664.1 | - | 12 | 1186 | |||
AK154499.1 | - | 21 | 2182 |