Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291146.1 | NP_001278075.1 | 2 | 478 | ||
NM_001291147.1 | NP_001278076.1 | 2 | 222 | |||
NM_001291148.1 | NP_001278077.1 | 2 | 184 | |||
NM_033562.4 | NP_291040.1 | 2 | 187 | |||
GenBank mRNA | AF208064.1 | - | 2 | 126 | ||
AK048627.1 | - | 2 | 134 | |||
AK077659.1 | - | 2 | 138 | |||
AK082564.1 | - | 2 | 147 | |||
AK083632.1 | - | 2 | 126 | |||
AK083725.1 | - | 1 | 896 | |||
AK090105.1 | - | 2 | 197 | |||
AK151934.1 | - | 2 | 478 | |||
BC005682.1 | - | 2 | 126 | 3154448 | ||
BE627680.1 | - | 2 | 155 |