Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276455.1 | NP_001263384.1 | 6 | 1525 | ||
NM_054087.3 | NP_473428.1 | 6 | 1639 | |||
GenBank mRNA | AF179403.1 | - | 6 | 1630 | ||
AF216204.1 | - | 6 | 1609 | |||
AF326916.1 | - | 6 | 1318 | |||
AF360396.1 | - | 6 | 1621 | |||
AF418986.1 | - | 6 | 1516 | |||
AK044825.1 | - | 6 | 1640 | |||
BC004021.1 | - | 2 | 193 | |||
BC034659.1 | - | 6 | 1620 | 5359902 |