Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054094.5 | NP_473435.1 | 14 | 1740 | ||
GenBank mRNA | AB059428.1 | - | 14 | 1744 | ||
AK149550.1 | - | 14 | 1750 | |||
AK149586.1 | - | 13 | 1685 | |||
BC016414.1 | - | 14 | 1737 | 4238964 | ||
BC022149.1 | - | 14 | 1718 | 5102652 |