Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080445.4 | NP_536693.1 | 1 | 865 | ||
GenBank mRNA | AK033608.1 | - | 1 | 775 | ||
AK078083.1 | - | 1 | 837 | |||
AK090344.1 | - | 1 | 807 | |||
AY050569.1 | - | 1 | 809 | |||
BC057933.1 | - | 1 | 817 | |||
BC082998.1 | - | 1 | 812 | 5011578 |