Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078477.2 | NP_510962.2 | 2 | 1655 | ||
GenBank mRNA | AF283891.1 | - | 2 | 1653 | ||
AK154524.1 | - | 2 | 1682 | |||
AK167841.1 | - | 1 | 566 | |||
AK167940.1 | - | 1 | 566 | |||
BC021615.1 | - | 1 | 88 | |||
BC038918.1 | - | 1 | 100 | |||
BC137607.1 | - | 2 | 1588 | 8860626 | ||
BC137609.1 | - | 2 | 1588 | 8860628 |