Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289564.1 | NP_001276493.1 | 15 | 1758 | ||
NM_001289566.1 | NP_001276495.1 | 15 | 1800 | |||
NM_080468.2 | NP_536716.2 | 16 | 1830 | |||
GenBank mRNA | AF346501.1 | - | 16 | 1830 | ||
AK034592.1 | - | 16 | 1743 | |||
AK047861.1 | - | 2 | 893 | |||
BC120741.1 | - | 16 | 1799 | 40129664 | ||
BC144802.1 | - | 16 | 1727 | 9053350 | ||
BC144803.1 | - | 16 | 1769 | 9053351 |