Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207223.1 | NP_997106.1 | 1, 2 | 576 | ||
GenBank mRNA | AK147505.1 | - | 1, 2 | 630 | ||
BC025556.1 | - | 1, 2 | 150 | |||
BC067016.1 | - | 1, 2 | 576 | 6827624 | ||
BC098196.1 | - | 1, 2 | 152 | 6847993 |