Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207223.1 | NP_997106.1 | 17 | 1961 | ||
GenBank mRNA | AK145262.1 | - | 11 | 915 | ||
AK147505.1 | - | 17 | 2015 | |||
AK159055.1 | - | 14 | 1594 | |||
AK185907.1 | - | 3 | 315 | |||
BC025556.1 | - | 17 | 1534 | |||
BC067016.1 | - | 17 | 1961 | 6827624 | ||
BC098196.1 | - | 17 | 1537 | 6847993 |