Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255993.1 | NP_001242922.1 | 5 | 1388 | ||
NM_001255994.1 | NP_001242923.1 | 4 | 1241 | |||
NM_080562.5 | NP_542129.2 | 4 | 1246 | |||
GenBank mRNA | AF360997.1 | - | 4 | 1176 | ||
AK034114.1 | - | 4 | 1195 | |||
AK049276.1 | - | 3 | 1012 | |||
AK158052.1 | - | 5 | 1346 | |||
AK170024.1 | - | 5 | 3089 | |||
AK170100.1 | - | 4 | 1218 | |||
BC025068.1 | - | 3 | 1108 | 4242459 |