Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025312.1 | NP_001020483.1 | 3 | 761 | ||
NM_133865.2 | NP_598626.2 | 4 | 927 | |||
GenBank mRNA | AK049115.1 | - | 4 | 922 | ||
AK051104.1 | - | 4 | 882 | |||
AK084347.1 | - | 3 | 756 | |||
AK134324.1 | - | 4 | 687 | |||
AK137495.1 | - | 4 | 484 | |||
AK138568.1 | - | 4 | 985 | |||
AK154124.1 | - | 4 | 829 | |||
BC011094.1 | - | 3 | 386 | 3493038 | ||
BC067017.1 | - | 3 | 670 | 6827667 | ||
BC125277.1 | - | 3 | 382 | |||
BC144725.1 | - | 3 | 372 |