Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133774.4 | NP_598535.1 | 2 | 227 | ||
GenBank mRNA | AF480297.1 | - | 2 | 82 | ||
AF480298.1 | - | 2 | 82 | |||
AK042347.1 | - | 2 | 231 | |||
AK042691.1 | - | 2 | 187 | |||
AK054501.1 | - | 2 | 228 | |||
AK078858.1 | - | 2 | 231 | |||
AK085331.1 | - | 2 | 231 | |||
AK208141.1 | - | 2 | 139 | |||
BC005642.1 | - | 2 | 227 | 3709232 |