Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130879.2 | NP_570949.2 | 16, 17 | 2283 | ||
GenBank mRNA | AK052297.1 | - | 16, 17 | 2225 | ||
AK133251.1 | - | 17, 18 | 2300 | |||
BC058410.1 | - | 3, 4 | 326 |