Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133223.4 | NP_573486.1 | 3, 4 | 357 | ||
GenBank mRNA | AB040819.1 | - | 3, 4 | 224 | ||
AK089930.1 | - | 3, 4 | 358 | |||
AK150203.1 | - | 3, 4 | 362 | |||
AY974601.1 | - | 2, 3 | 196 | |||
BC025842.1 | - | 2, 3 | 229 | |||
BC117028.1 | - | 3, 4 | 355 | 40126347 | ||
BC119041.1 | - | 3, 4 | 355 | 8733790 |