Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133245.1 | NP_573508.1 | 207 | |||
GenBank mRNA | AF060220.1 | - | 191 | |||
AF364516.1 | - | 190 | ||||
AK011084.1 | - | 207 | ||||
AK028172.1 | - | 207 | ||||
AK146724.1 | - | 201 | ||||
BC110627.2 | - | 185 | 40051901 |