Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167991.1 | NP_001161463.1 | 8 | 644 | ||
NM_133255.2 | NP_573518.2 | 8 | 644 | |||
GenBank mRNA | AK030599.1 | - | 8 | 637 | ||
AK042048.1 | - | 8 | 632 | |||
AK136069.1 | - | 8 | 638 | |||
AK145768.1 | - | 8 | 612 | |||
AK161612.1 | - | 8 | 597 | |||
AK166954.1 | - | 8 | 624 | |||
AK169073.1 | - | 8 | 612 | |||
AK191641.1 | - | 4 | 184 | |||
BC019486.1 | - | 8 | 620 | 4211990 | ||
BC029967.1 | - | 8 | 585 | |||
BC055881.1 | - | 8 | 590 | 3590336 | ||
BC080744.1 | - | 8 | 634 | 30471369 |