Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134185.2 | NP_598946.2 | 1 | 468 | ||
GenBank mRNA | BC132241.1 | - | 1 | 468 | 40130518 | |
BC132577.1 | - | 1 | 468 | 40130854 |
Gene Symbol: Vmn1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053231.2 | NP_444461.2 | 1 | 421 |
Gene Symbol: Vmn1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134187.3 | NP_598948.1 | 1 | 478 | ||
GenBank mRNA | AF394951.1 | - | 1 | 478 |