Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167535.1 | NP_001161007.1 | 1 | 176 | ||
GenBank mRNA | BC145887.1 | - | 1 | 229 | 40131190 |
Gene Symbol: Vmn1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167534.1 | NP_001161006.1 | 1 | 176 |
Gene Symbol: Vmn1r238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167539.1 | NP_001161011.1 | 1 | 176 |
Gene Symbol: Vmn1r79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166835.1 | NP_001160307.1 | 1 | 176 |