Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138682.2 | NP_619623.2 | 1 | 2034 | ||
GenBank mRNA | AF300458.1 | - | 1 | 1991 | ||
AK046781.1 | - | 1 | 1241 | |||
BC012209.1 | - | 1 | 2035 | |||
BC117833.1 | - | 1 | 1975 | 40092947 | ||
BC117834.1 | - | 1 | 1975 | 40092948 | ||
DQ177325.1 | - | 1 | 2034 |