Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081047.1 | NP_001074516.1 | 19 | 1675 | ||
GenBank mRNA | AK145910.1 | - | 19 | 1637 | ||
BC113805.1 | - | 19 | 1640 | 40062208 | ||
BC113956.1 | - | 19 | 1640 | 40062209 |