Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183138.2 | NP_898961.2 | 1, 2 | 140 | ||
GenBank mRNA | AK044758.1 | - | 1, 2 | 93 | ||
AK046543.1 | - | 1, 2 | 141 | |||
AK046552.1 | - | 1, 2 | 141 | |||
AK046553.1 | - | 1, 2 | 141 | |||
HQ423151.1 | - | 1, 2 | 314 | |||
JX946278.1 | - | 2, 3 | 846 |