Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085414.2 | NP_001078883.2 | 3, 4 | 961 |
Gene Symbol: Gm13078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085412.2 | NP_001078881.1 | 3, 4 | 969 |