Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002596.3 | NP_002587.2 | 6 | 837 | ||
NM_212502.2 | NP_997667.1 | 6 | 755 | |||
NM_212503.2 | NP_997668.1 | 6 | 845 | |||
XM_011509602.2 | XP_011507904.1 | 6 | 895 | |||
XM_017001423.1 | XP_016856912.1 | 6 | 990 | |||
GenBank mRNA | AB209307.1 | - | 4 | 1417 | ||
AK126879.1 | - | 3 | 841 | |||
AK293756.1 | - | 5 | 1119 | |||
AK295272.1 | - | 4 | 285 | |||
AK295654.1 | - | 5 | 900 | |||
AK296320.1 | - | 6 | 803 | |||
AK308913.1 | - | 4 | 286 | |||
AU130437.1 | - | 6 | 716 | |||
AY353237.1 | - | 6 | 670 | |||
AY353238.1 | - | 6 | 734 | |||
X66362.1 | - | 3 | 211 |