Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312673.1 | NP_001299602.1 | 8 | 885 | ||
NM_005017.3 | NP_005008.2 | 9 | 949 | |||
GenBank mRNA | AK304534.1 | - | 8 | 742 | ||
AK313450.1 | - | 8 | 807 | |||
BC046355.1 | - | 9 | 910 | 5744747 | ||
L28957.1 | - | 8 | 782 |