Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039388.2 | NP_001034477.1 | 2 | 353 | ||
NM_001039389.2 | NP_001034478.1 | 2 | 274 | |||
NM_172445.3 | NP_766033.1 | 2 | 274 | |||
GenBank mRNA | AK014128.1 | - | 2 | 250 | ||
AK032582.1 | - | 2 | 236 | |||
AK036203.1 | - | 2 | 270 | |||
AK044740.1 | - | 2 | 254 | |||
AK129254.1 | - | 2 | 230 | |||
AK146209.1 | - | 2 | 249 | |||
BB611667.1 | - | 2 | 353 | |||
BC046236.1 | - | 1 | 101 | 6817089 | ||
BC048819.1 | - | 2 | 198 | |||
CJ134699.1 | - | 2 | 275 |